missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Granular, U.S.P., J.T. Baker™
Specifications
Specifications
| Physical Form | Granules |
| Quantity | 500 g |
| Formula Weight | 98.14 |
| Percent Purity | 100.0% |
| Grade | USP |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.57kg |
| Chemical Name or Material | Potassium Acetate |