Learn More
Potassium Acetate (Crystalline/USP/EP/BP), Fisher Chemical™
Supplier: Fisher Chemical P18112
Specifications
| Potassium Acetate | |
| 127-08-2 | |
| 7.0 to 8.0 | |
| 12 kg | |
| CH3COOK | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| 99.0 to 101.0% (BP) | |
| Pass Test (BP), Pass Test (EP), Pass Test (USP), Pass Test (USP/EP/BP) | |
| Poly Pail |
| 292°C | |
| White | |
| Solid | |
| C2H3KO2 | |
| MFCD00012458 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.14 | |
| BP/EP/USP | |
| 1.0% max. (USP), 1.0% max. (USP), 3.0% max. (EP/BP), 3.0% max. (BP), 3.0% max. (EP) |
Chemical Identifiers
| 127-08-2 | |
| 98.142 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0