missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate (Crystalline/USP/EP/BP), Fisher Chemical™

Catalog No. p-46761
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P181212 2.5 kg Poly Bottle
P18112 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P181212 Supplier Thermo Fisher Scientific Supplier No. P181212
Only null left

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Color White
pH 7.0 to 8.0
Physical Form Solid
Quantity 2.5 kg
Linear Formula CH3COOK
Formula Weight 98.14
Percent Purity 99.0 to 101.0% (BP)
Grade BP/EP/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (USP), Pass Test (USP/EP/BP)
Loss on Drying 1.0% max. (USP), 1.0% max. (USP), 3.0% max. (EP/BP), 3.0% max. (BP), 3.0% max. (EP)
Packaging Poly Bottle
Chemical Name or Material Potassium Acetate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0