missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical
PO155, 127-08-2, C2H3KO2
$303.33 - $9196.30
Chemical Identifiers
| CAS | 127-08-2 |
|---|---|
| Molecular Formula | C2H3KO2 |
| Molecular Weight (g/mol) | 98.14 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| IUPAC Name | potassium acetate |
| SMILES | [K+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605429
|
Spectrum Chemical Mfg Cor
PO155125GM |
125 g | Poly Bottle |
Each for $303.33
|
|
||||
|
18605430
|
Spectrum Chemical Mfg Cor
PO155500GM |
500 g | Poly Bottle |
Each for $797.53
|
|
||||
|
18605432
|
Spectrum Chemical Mfg Cor
PO15525KG |
2.5 kg | Poly Pail |
Each for $2,715.90
|
|
||||
|
18605431
|
Spectrum Chemical Mfg Cor
PO155500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $4,126.40
|
|
||||
|
18605433
|
Spectrum Chemical Mfg Cor
PO15525KGCS4 |
4 x 2.5 kg | Poly Pail |
Case of 4 Each for $9,196.30
|
|
||||
|
18605434
|
Spectrum Chemical Mfg Cor
PO15512KG |
12 kg | Poly Pail |
Each for $4,468.60
|
|
||||
|
18605435
|
Spectrum Chemical Mfg Cor
PO15550KGBL |
50 kg | Fiber Drum |
Each for $9,149.37
|
|
||||