missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical

PO155, 127-08-2, C2H3KO2

Supplier:  Spectrum Chemical Mfg Cor PO155500GMCS6

 View more versions of this product

Catalog No. 18605431

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Acetate, Crystalline Powder, USP is a potassium salt used to replenish electrolytes, for restoration of water-electrolyte balance, as well as, a urinary and systemic alkalizer. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

127-08-2
7.5 to 8.5
C2H3KO2
[K+].CC([O-])=O
98.14
0.01
1
6 x 500 g
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate
99 to 100.5%
Poly Bottle

Chemical Identifiers

127-08-2
98.14
potassium acetate
C2H3KO2
SCVFZCLFOSHCOH-UHFFFAOYSA-M
[K+].CC([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions