missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
$93.40 - $152.18
Chemical Identifiers
| CAS | 127-08-2 |
|---|---|
| Molecular Formula | C2H3KO2 |
| Molecular Weight (g/mol) | 98.142 |
| MDL Number | MFCD00012458 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
| PubChem CID | 517044 |
| ChEBI | CHEBI:32029 |
| IUPAC Name | potassium;acetate |
| SMILES | CC(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62817AE
|
Thermo Scientific Chemicals
J62817AE |
100 mL |
Each for $93.40
|
|
|||||
|
AAJ62817AK
|
Thermo Scientific Chemicals
J62817AK |
250 mL |
Each for $152.18
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 127-08-2 | |
| 98.142 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
Specifications
| Colorless | |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| 7.5 | |
| Liquid |
| 127-08-2 | |
| CH3CO2K | |
| 1M aq. soln. | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.15 | |
| 100 mL | |
| Potassium acetate |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures