missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium AA Standard, 1000 ppm K in H2O, Ricca Chemical
Supplier: Ricca Chemical Company AK1KW100
Specifications
| Potassium AA Standard | |
| 7447-40-7,7732-18-5 | |
| 100.0,0.19 | |
| ClK | |
| Colorless | |
| 100°C | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 100 mL | |
| CHEBI:32588 |
| Natural LDPE Bottle | |
| 98.0,0.19 | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| 1mL = 1mg K (1, 000ppm K)KCl in Water |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Safety and Handling
ShelfLife : 24 months