missing translation for 'onlineSavingsMsg'
Learn More

Potassium 7-Hydroxy-1-naphthalenesulfonate (contains isomer) 90.0+%, TCI America™

Catalog No. N007525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007525G 25 g
N0075500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N007525G Supplier TCI America Supplier No. N007525G
Only null left

Chemical Identifiers

CAS 30252-40-5
Molecular Formula C10H7KO4S
Molecular Weight (g/mol) 262.32
MDL Number MFCD00065327
InChI Key UZBFJXMJCARTGH-UHFFFAOYSA-M
PubChem CID 23663680
IUPAC Name potassium 7-hydroxynaphthalene-1-sulfonate
SMILES [K+].OC1=CC=C2C=CC=C(C2=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
Synonym Bayer′Cs Acid Potassium Salt, Croceic Acid Potassium Salt, 7-Hydroxy-1-naphthalenesulfonic Acid Potassium Salt, 2-Naphthol-8-sulfonic Acid Potassium Salt, Potassium 2-Naphthol-8-sulfonate
Formula Weight 262.32
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 7-Hydroxy-1-naphthalenesulfonate (contains isomer)