accessibility menu, dialog, popup

UserName

KHOSOOO

CAS RN 30252-40-5

IUPAC Name: potassium 7-hydroxynaphthalene-1-sulfonate
Synonyms: potassium 7-hydroxynaphthalene-1-sulfonate
Molecular Weight (g/mol): 262.32
Molecular Formula: C10H7KO4S
InChi Key: UZBFJXMJCARTGH-UHFFFAOYSA-M
SMILES: [K+].OC1=CC=C2C=CC=C(C2=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results