missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-iodophenyltrifluoroborate, 96%

Catalog No. AAH2875803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2875803 1 g
AAH2875806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2875803 Supplier Thermo Scientific Chemicals Supplier No. H2875803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 912350-00-6 | C6H4BF3IK | 309.906 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 912350-00-6
Molecular Formula C6H4BF3IK
Molecular Weight (g/mol) 309.906
MDL Number MFCD09800738
InChI Key WTXNSBOVCAXUGV-UHFFFAOYSA-N
Synonym potassium 4-iodophenyltrifluoroborate, potassium trifluoro 4-iodophenyl boranuide, amtb080, potassium trifluoro 4-iodophenyl borate 1-
PubChem CID 23688160
IUPAC Name potassium;trifluoro-(4-iodophenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)I)(F)(F)F.[K+]

Specifications

Melting Point 297°C to 299°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 309.9
Percent Purity 96%
Chemical Name or Material Potassium 4-iodophenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only