CAS RN 912350-00-6
IUPAC Name:
potassium trifluoro(4-iodophenyl)boranuide
Synonyms:
potassium trifluoro(4-iodophenyl)boranuide
Molecular Weight (g/mol):
309.91
Molecular Formula:
C6H4BF3IK
InChi Key:
WTXNSBOVCAXUGV-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC=C(I)C=C1
1
–
1
of
1
results
1
Promotions Available
Potassium 4-iodophenyltrifluoroborate, 96%
Potassium 4-iodophenyltrifluoroborate, 96%
CAS: 912350-00-6 Molecular Formula: C6H4BF3IK Molecular Weight (g/mol): 309.906 MDL Number: MFCD09800738 InChI Key: WTXNSBOVCAXUGV-UHFFFAOYSA-N Synonym: potassium 4-iodophenyltrifluoroborate,potassium trifluoro 4-iodophenyl boranuide,amtb080,potassium trifluoro 4-iodophenyl borate 1- PubChem CID: 23688160 IUPAC Name: potassium;trifluoro-(4-iodophenyl)boranuide SMILES: [B-](C1=CC=C(C=C1)I)(F)(F)F.[K+]