missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (2-Formylphenyl)trifluoroborate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 222°C |
| Quantity | 1 g |
| Formula Weight | 212.02 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Potassium (2-Formylphenyl)trifluoroborate |
Safety and Handling
| TSCA | No |