missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2,4-difluorophenyltrifluoroborate, 95%

Catalog No. AAH3732603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3732603 1 g
AAH3732606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3732603 Supplier Thermo Scientific Chemicals Supplier No. H3732603
Only null left

CAS: 871231-41-3 | C6H3BF5K | 219.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 871231-41-3
Molecular Formula C6H3BF5K
Molecular Weight (g/mol) 219.99
MDL Number MFCD07784375
InChI Key HZKKNPYRMYVEOW-UHFFFAOYSA-N
Synonym potassium 2,4-difluorophenyltrifluoroborate, potassium 2,4-difluorophenyl trifluoroborate, potassium 2,4-difluorophenyl trifluoroboranuide, pubchem11329, potassium 2,4-difluorophenyl trifluoro borate 1-
PubChem CID 23697340
IUPAC Name potassium;(2,4-difluorophenyl)-trifluoroboranuide
SMILES [B-](C1=C(C=C(C=C1)F)F)(F)(F)F.[K+]

Specifications

Quantity 1 g
Formula Weight 219.99
Percent Purity 95%
Chemical Name or Material Potassium 2,4-difluorophenyltrifluoroborate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only