missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2,4,5-Trichlorophenoxyacetate 98.0+%, TCI America™

Catalog No. p-7136810
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T150925G 25 g
T1509500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T150925G Supplier TCI America Supplier No. T150925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37785-57-2
Molecular Formula C8H4Cl3KO3
Molecular Weight (g/mol) 293.565
MDL Number MFCD00191590
InChI Key LUDUIQAQFKIYND-UHFFFAOYSA-M
Synonym 2,4,5-Trichlorophenoxyacetic Acid Potassium Salt, 2,4,5-T Potassium Salt
PubChem CID 23684557
IUPAC Name potassium;2-(2,4,5-trichlorophenoxy)acetate
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)OCC(=O)[O-].[K+]

Specifications

Color White
Quantity 25 g
Formula Weight 293.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 2,4,5-Trichlorophenoxyacetate