accessibility menu, dialog, popup

UserName

CAS RN 37785-57-2

IUPAC Name: potassium 2-(2,4,5-trichlorophenoxy)acetate
Synonyms: potassium 2,4,5-trichlorophenoxyacetate
Molecular Weight (g/mol): 293.57
Molecular Formula: C8H4Cl3KO3
InChi Key: LUDUIQAQFKIYND-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)COC1=CC(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results