missing translation for 'onlineSavingsMsg'
Learn More

Potash Sulfurated, Technical, Approx. 12%, Spectrum™ Chemical

$2083.36 - $2083.36

Chemical Identifiers

CAS 39365-88-3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612434
View Documents
Spectrum Chemical Mfg Cor
P116025KG
2.5 kg Amber Glass Bottle
Each for $2,083.36
Only null left
 
Description

Description

Specifications

Chemical Identifiers

39365-88-3

Specifications

39365-88-3
2.5 kg
Technical
1
∼12%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.