accessibility menu, dialog, popup

UserName

OSOSOSKKKK

CAS RN 39365-88-3

IUPAC Name: tetrapotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate) sulfanediide
Synonyms: tetrapotassium Hypo sulfanediide
Molecular Weight (g/mol): 300.57
Molecular Formula: K4O3S3
InChi Key: YFKFGPGTVQHNEF-UHFFFAOYSA-L
SMILES: [S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)