missing translation for 'onlineSavingsMsg'
Learn More

Potash Sulfurated, Technical, Approx. 12%, Spectrum™ Chemical

Catalog No. 18612434
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612434 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18612434 Supplier Spectrum Chemical Mfg Cor Supplier No. P11602.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39365-88-3

Specifications

CAS Max % 1
Physical Form Irregular Lumps/Pieces
Quantity 2.5 kg
Percent Purity ∼12%
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Potash Sulfurated

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.