Learn More
Thermo Scientific™ POROS™ GoPure™ Caprylate Pre-packed Column, 0.5 x 5 cm, 1 mL
Thermo Scientific™ POROS™ GoPure™ pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.
Supplier: Thermo Scientific™ A51054
Description
POROS GoPure pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.
These pre-packed columns are ready to connect and have the following features:
- Packed with POROS chromatography resins that offer superior performance over a wide range of flow rates
- 1-mL and 5-mL formats provide a uniform screening platform for process development
- Convenient, ready-to-run columns that simplify and speed up POROS resin evaluations and enable small scale purification
- Direct connection to standard chromatography systems using 1/16 in., 10-32 fittings
POROS Caprylate Mixed-Mode Chromatography Resin (Cat. No. A51051) is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species.
Features of POROS Caprylate Mixed-Mode resin include:
- Selective capture of aggregates and hydrophobic high-molecular-weight impurities
- Flow-through design that helps save buffer and material costs
- Excellent scalability and high productivity
- Wide operational range
- Non-animal-derived
POROS Caprylate Mixed-Mode resin operates in flow-through mode which helps reduce costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems. POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
- Average particle size: 50 μm
- Ligand: octanoic acid
- Binding capacity: >20 g lysozyme/L resin
- Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
- Clean-in-place: 0.5 M NaOH
- Formulation buffer: 0.1 M NaOH
- Mechanical resistance: 100 bar (10 MPa, 1450 psi)
Bulk resin is available and these columns come with a regulatory support package. Worldwide technical support is also available.
Other POROS chromatography resins are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
- Environmental benefits include:
- Recyclable
Specifications
1 mL | |
50 μm | |
1 column | |
Pre-packed Column | |
Chromatography Column | |
Process Development |
Store at 2–30°C. |
|
POROS™ | |
Monoclonal Antibodies | |
Caprylic Acid | |
50 mm |
For Research Use Only. Not for use in diagnostic procedures.