missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ Ethyl Hydrophobic Interaction Chromatography (HIC) Resin
GREENER_CHOICE

Catalog No. A32557
Encompass
Change view
Click to view available options
Quantity:
25 mL
50 mL
250 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32557 25 mL
A32556 50 mL
361054718 250 mL
A32554 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A32557 Supplier Thermo Scientific™ Supplier No. A32557

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ POROS™ Ethyl Hydrophobic Interaction Chromatography (HIC) Resin is a bioprocess high-performance resin engineered for improved impurity removal during large-scale downstream purification of biomolecules.

POROS Ethyl Hydrophobic Interaction Chromatography (HIC) Resin is a bioprocess high-performance resin engineered for improved impurity removal during large-scale downstream purification of biomolecules. This unique resin provides high resolution and capacity while maintaining excellent pressure flow characteristics, helping increase productivity.

POROS Ethyl HIC Resin is designed on the well-established cross‐linked poly(styrene-divinylbenzene) POROS base bead with an attached novel hydrophobic ethyl ligand, a resin that is primarily suited for bind/elute purifications.

Features of POROS Ethyl HIC Resin include:

  • Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50 micron particle size. Higher flow rates help improve productivity.
  • Unique selectivity due to the novel hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
  • High capacity, reducing the process volume significantly for subsequent steps
  • A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process

These features of POROS Ethyl HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography. POROS Ethyl HIC Resin combines superior and consistent performance, exceptional customer support, strong operational excellence, and a strong legacy based on proven technology from a well-established chromatography resin supplier. This resin comes with complete technical and regulatory support. POROS Ethyl HIC Resin is available in various bulk package sizes for large-scale purification, as well as pre-packed columns for initial process development.

Main characteristics

  • Support matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: novel ethyl
  • pH range: 1–14
  • Ionic strength range: 0 to 5 M, all common salts
  • Mechanical resistance: 100 bar (1450 psi; 10 MPa)
  • Shipping solution: 18% (v/v) ethanol

Specifications

Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Content And Storage Store at 2°C to 30°C. Do not freeze.
Shipping Condition Ambient
Particle Size 50 μm
Form Resin Slurry
Product Line POROS
Quantity 25 mL
Type Resin
Stationary Phase Novel Ethyl