missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ LC Lambda Affinty HPLC Columns

Catalog No. p-200156847
Change view
Click to view available options
Dimensions:
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
4469150 2.1 x 30 mm
4469163 4.6 x 50 mm
4469168 4.6 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4469150 Supplier Thermo Scientific™ Supplier No. 4469150
Only null left

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates.

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

POROS CaptureSelect LC Lambda HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three minute quantitation of Fabs, immunoglobulins, and bispecific antibodies having a Lambda light chain.

Features:

  • Broad utility—POROS A, G and CaptureSelectaffinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—deliver high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect LC Lambda affinity ligands when combined with POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of all immunoglobulins and Fabs having a Lambda light chain (all subclasses).

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

LC Lambda Column Specifications:

  • Protein Binding Capacity: 10 mg/mL
  • Species Specificity: Human
  • Column Material: PEEK™ polymer or stainless steel
  • Frit: 2 μm, titanium
  • POROS Resin: Cross-linked poly(styrenedivinylbenzene)
  • CaptureSelect Ligands: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae

Specifications

Column Format Analytical Column, Biocompatible PEEK
Diameter (Metric) 2.1 mm
Flow Rate Up to 5000 cm/hr.
For Use With HPLC
Length (Metric) 30 mm
Max. Pressure 2400 psi (170 bar)
Particle Size 20 μm
pH 1-10
Pore Size 2000 Å
Quantity 1 Column
Temperature 2-40°C
Stationary Phase Light Chain Lambda affinity ligand, human-specific
Content And Storage Column includes EZ grip fittings and column test certificate

Store at 4°C. DO NOT FREEZE.
Product Line POROS CaptureSelect
Binding Capacity 10.0 mg/mL IgG
Elution Volume Max 200 μL
Elution Volume Min 100 μL
Reactivity Human
High-throughput Compatibility Not High-throughput Compatible (Manual)
Sample Type Cell Culture Supernatants
Target Molecule Lambda Light Chain
Shipping Condition Wet Ice
Matrix Polystyrenedivinylbenzene
Volume (Metric) 100 μL
Dimensions 2.1 x 30 mm
Packing Material Highly Crosslinked Styrene-Divinylbenzene polymer (ST-DVB)
Column Type Affinity
Material PEEK
Technique Bio-LC
Show More Show Less

For Research Use Only. Not for use in diagnostics procedures.