missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ HSA Affinty HPLC Columns

Catalog No. 4469170
Change view
Click to view available options
Dimensions:
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
4469170 4.6 x 100 mm
4469151 2.1 x 30 mm
4469165 4.6 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4469170 Supplier Thermo Scientific™ Supplier No. 4469170

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates.

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

POROS CaptureSelect HSA HPLC columns incorporate the advantages of high-speed Poros chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three minute quantitation of HSA fusion proteins.

Features:

  • Broad utility—POROS A, G and CaptureSelectaffinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—deliver high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect HSA affinity ligands when combined with POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of HSA Fusion proteins.

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

HSA Column Specifications:

  • Protein Binding Capacity: 2.6 mg/mL
  • Species Specificity: Human
  • Column Material: PEEK™ polymer or stainless steel
  • Frit: 2 μm, titanium
  • POROS Resin: Cross-linked poly (styrenedivinylbenzene)
  • CaptureSelect Ligands: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae

Specifications

Flow Rate Up to 5000 cm/hr.
Length (Metric) 100 mm
Max. Pressure 180 bar
Particle Size 20 μm
Pore Size 2000 Å
Quantity 1 Column
Stationary Phase Human Serum Albumin affinity ligand, humanspecific
Content And Storage Column includes EZ grip fittings and column test certificate

Store at 4°C. DO NOT FREEZE.
Product Line CaptureSelect, POROS
Binding Capacity 2.6 mg/mL
Elution Volume Max 3.4 mL
Elution Volume Min 1.7 mL
Reactivity Human
High-throughput Compatibility Not High-throughput Compatible (Manual)
Sample Type Cell Culture Supernatants
Target Molecule HSA, HSA Fusion Proteins
Shipping Condition Wet Ice
Matrix Polystyrenedivinylbenzene
Volume (Metric) 1.7 mL
Dimensions 4.6 x 100 mm
Column Type Affinity
Material PEEK
Type Column
Purification Target HSA, HSA Fusion Proteins
Show More Show Less

For Research Use Only. Not for use in diagnostics procedures.