missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ GCSF Affinty HPLC Columns

Catalog No. p-200156840
Change view
Click to view available options
Dimensions:
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
4485157 2.1 x 30 mm
4485164 4.6 x 50 mm
4485168 4.6 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4485157 Supplier Thermo Scientific™ Supplier No. 4485157
Only null left

Thermo Scientific™ POROS™ CaptureSelect™ GCSF HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands.

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentations broths, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle. POROS CaptureSelect GCSF HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three-minute quantitation of granulocyte colony-stimulating factor (GCSF).

  • Broad utility—POROS A, G, and CaptureSelect affinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins, fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—allows for high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect GCSF affinity ligand and POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid titer determination and small-scale purification of GCSF.

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and allows for an eluted product that is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows for the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime can result in strong re-use performance. POROS affinity chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

Specifications

Length (Metric) 30 mm
Max. Pressure 2600 psi, 180 bar
Particle Size 20 μm
Pore Size 2000 Å
Quantity 1 Column
Stationary Phase Granulocyte ColonyStimulating Factor affinity ligand, human-specific
Content And Storage Column includes EZ grip fittings and column test certificate.
Store at 2°C to 8°C. DO NOT FREEZE.
Product Line CaptureSelect, POROS
Binding Capacity 5.2 mg/mL (ligand density)
Elution Volume Max 29 column volumes
Elution Volume Min 14 column volumes
High-throughput Compatibility Automated Protocols
Target Molecule Protein
Matrix POROS
Volume (Metric) 0.1 mL
Dimensions 2.1 x 30 mm
Column Type Affinity
Material PEEK
Type Column
Purification Target Protein
For Use With (Application) FPLC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.