Learn More
Thermo Scientific™ POROS™ CaptureSelect™ FcXL Affinity HPLC Columns
Description
POROS CaptureSelect FcXL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect™ affinity ligands.
CaptureSelect FcXL Affinity HPLC columns recognize the CH3 domain of human IgG antibodies, enabling quantitation and small-scale purification of all IgG subclasses (IgG1, IgG2, IgG3, and IgG4), as well as Fab fragments and fusion proteins containing the intact CH3 domain, independent of light-chain iso-type and source material.
Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.
Features of POROS CaptureSelect FcXL Affinity HPLC columns include:
- Versitile—applicable for IgG Fab fragmetns, all human IgG subclasses, and IgG fusion proteins with intact CH3 domain
- Specific—no cross-binding to bovine, horse, or rodent IgG
- Fast—POROS resins enable quantitation or purification of immunoglobulins and fusion proteins in minutes
- Hands-off operation—HPLC method can be automated for a streamlined operation
- Robust—designed to deliver high-performance purification over hundreds of chromatography cycles
High speed with high specificity
POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis, including mass spectrometry.
Automation
POROS Affinity HPLC columns are for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligand enables one method and buffer system to be used across a class of molecules, streamlining analytical approaches.
Multiple cycles
Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regimen results in strong re-use performance. POROS Affinity HPLC columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.
Main characteristics
- Protein binding capacity: 7.3 mg/mL
- Species specificity: human
- Column material: PEEK polymer
- Frit: 2 μm, titanium
- POROS resin: cross-linked poly(styrene-divinylbenzene)
- CaptureSelect ligands: single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae
Specifications
Specifications
| Column Format | Analytical Column, Biocompatible PEEK |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 30 mm |
| Max. Pressure | 2400 psi (170 bar) |
| Particle Size | 20 μm |
| pH | 1-10 |
| Pore Size | 2000 Å |
| Quantity | 1 Column |
| Temperature | 2-40°C |
| Stationary Phase | IgG Fc affinity ligand, human‐specific |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.