Learn More
Eisco™ Polystyrene Petri Dishes
Disposable, sterile polystyrene petri dishes ideal for microbiology applications.
Supplier:  Eisco™ CH2020011
 View more versions of this product
Catalog No. S15093
Only null left
Explore available promotions
Description
Description
  • Made of high quality polystyrene
  • Built-in triple vents aid gaseous exchange
  • Multifunctional and ready-to-use
  • Clear and shatter resistant
  • Sterilized in package
Specifications
Specifications
Not Autoclavable
Clear
Not Gridded
Round
Non-treated
1
1 Compartment
15 mm
Flat
Limited-use
Polystyrene
Sterile
Petri Dish
Cell Culture
Triple
100 mm
Product Suggestions
Product Suggestions