missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80, Multi-Compendial, N.F., J.T. Baker™
Supplier: Avantor 411707
| Quantity | 19 L |
|---|---|
| Packaging | Steel Drum |
Chemical Identifiers
| 9005-65-6 | |
| 604.82 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| MFCD00082107 | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Specifications
| Polysorbate 80 | |
| 19 L | |
| Steel Drum | |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
| Multi-Compendial/NF |
| 9005-65-6 | |
| 148.00°C | |
| 1L = 1.08kg | |
| MFCD00082107 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 604.82 |