missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80, Multi-Compendial, N.F., J.T. Baker™

Catalog No. p-5391658
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Steel Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003656 20 mL Narrow Mouth Amber Glass Bottle
02003654 200 mL Narrow Mouth Amber Glass Bottle
02003658 1 L Narrow Mouth Amber Glass Bottle
02003659 1 L Narrow Mouth Amber Glass Bottle, w/ Septum
02003662 4 L Narrow Mouth Amber Glass Bottle
02003657 19 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02003656 Supplier Avantor Supplier No. 411702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 20 mL
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.08kg
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 80