missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 20, MP Biomedicals™
$89.44 - $578.01
Chemical Identifiers
| CAS | 9005-64-5 |
|---|---|
| Molecular Formula | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165986 |
| InChI Key | HMFKFHLTUCJZJO-UHFFFAOYNA-N |
| Synonym | Polyoxyethylenesorbitan monolaurate, Tween 20 |
| PubChem CID | 443314 |
| SMILES | CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19484180
|
MP Biomedicals Inc
0219484180 |
100 mL |
Each for $89.44
|
|
|||||
|
ICN19472404
|
MP Biomedicals Inc
0219472404 |
4 L |
Each for $578.01
|
|
|||||
Chemical Identifiers
| 9005-64-5 | |
| 522.68 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| 443314 |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| MFCD00165986 | |
| Polyoxyethylenesorbitan monolaurate, Tween 20 | |
| CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Specifications
| Yellow | |
| Oily Liquid | |
| 100 mL | |
| MFCD00165986 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
| 443314 |
| 9005-64-5 | |
| 1.095 g/mL | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| Polyoxyethylenesorbitan monolaurate, Tween 20 | |
| CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Polysorbate 20 |