Learn More
Polysorbate 20, MP Biomedicals™
Supplier:  MP Biomedicals Inc 0219472404
 View more versions of this product
Catalog No. ICN19472404
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Polysorbate 20
9005-64-5
1.095 g/mL
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Polyoxyethylenesorbitan monolaurate, Tween 20
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Yellow
Oily Liquid
4 L
MFCD00165986
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
443314
Chemical Identifiers
9005-64-5
522.68
HMFKFHLTUCJZJO-UHFFFAOYNA-N
443314
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MFCD00165986
Polyoxyethylenesorbitan monolaurate, Tween 20
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Product Suggestions
Product Suggestions