Learn More
MilliporeSigma™ Supelco™ Polypropylene Screw Top Vials, Natural
Polypropylene vials offer an alternative to glass vials which are prone to breakage. These vials are well suited for storing pH sensitive samples, and eliminate the need for an insert when microsampling.
Supplier: MilliporeSigma™ Supelco™ 24710
Description
- Low extractables
- Compatible with many solvents
- Excellent choice when working with proteins
- Ideal for ion chromatography applications
Chemical Identifiers
| Natural | |
| Polypropylene | |
| Screw Thread Vial |
| 12 mm | |
| 32 mm |
Specifications
| Natural | |
| Polypropylene | |
| 1000/Pk. | |
| Screw Thread Vial |
| 12 mm | |
| 0.75 mL | |
| 32 mm |