missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Polypropylene, Carboy with Spigot
For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White
$287.25 - $406.81
Specifications
| Accuracy | ±5% |
|---|---|
| Autoclavable | Autoclavable |
| Closure Material | Polypropylene |
| Closure Size | 83 mm |
| Color | White |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
19-830-439
|
Justrite™
JMC12938 |
5 L |
Each for $287.25
|
|
|||||
|
19-830-440
|
Justrite™
JMC12939 |
10 L |
Each for $406.81
|
|
|||||
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
| ±5% | |
| Polypropylene | |
| White | |
| Spigot | |
| Carboy with Spigot |
| Autoclavable | |
| 83 mm | |
| For storing and dispensing solvents, buffers, reagents and powders. | |
| Polypropylene | |
| Rectangular |