missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Monododecyl Ether (n=approx. 25), TCI America™
Supplier: TCI America P0671500G
Specifications
| White-Yellow | |
| Crystal at 20°C | |
| C14H30O2 | |
| Polyethylene Glycol Monolauryl Ether | |
| SFNALCNOMXIBKG-UHFFFAOYSA-N | |
| 2-(dodecyloxy)ethan-1-ol |
| Polyethylene Glycol Monododecyl Ether (n=approx. 25) | |
| 9002-92-0 | |
| MFCD00043063 | |
| 500 g | |
| CCCCCCCCCCCCOCCO | |
| 230.39 |
Chemical Identifiers
| 9002-92-0 | |
| 230.39 | |
| SFNALCNOMXIBKG-UHFFFAOYSA-N | |
| 2-(dodecyloxy)ethan-1-ol |
| C14H30O2 | |
| MFCD00043063 | |
| Polyethylene Glycol Monolauryl Ether | |
| CCCCCCCCCCCCOCCO |
Safety and Handling
EINECSNumber : (7)-0097
RTECSNumber : JR5990000
TSCA : Yes