missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PEEK Sample Loops
Description
- Wetted parts are resistant to organic solvents
- Wetted parts are resistant to salt buffers
- Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polyetheretherketone (PEEK)
Specifications
Specifications
| Type | Sample Loop |
| Material | PEEK |
| Connection Size (Metric) | 1.58 mm |
| Diameter (Metric) Inner | 0.75 mm |
| Volume (Metric) | 2 mL |