missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 95.5-96.5% hydrolyzed, M.W. approx. 85,000-124,000

Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

Supplier:  Thermo Scientific Chemicals 183390025

 View more versions of this product

Catalog No. AC183390025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Poly(vinyl alcohol)
230.0°C to 240.0°C
Authentic
[-CH2CH(OH)-]n
2.5 kg
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
IMROMDMJAWUWLK-UHFFFAOYSA-N
ethenol
11199
95.5-96.5%
9002-89-5
4.5 to 6.5 (4% aq. soln.)
(C2H4O)n
MFCD00081922
15,7706
Solubility in water: soluble in hot water.
OC(-*)C-*
44.05
27-31 mPa.s (20°C 4% aq. sol.)

Chemical Identifiers

9002-89-5
44.05
IMROMDMJAWUWLK-UHFFFAOYSA-N
11199
OC(-*)C-*
(C2H4O)n
MFCD00081922
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
ethenol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only