missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 95.5-96.5% hydrolyzed, M.W. approx. 85,000-124,000

Catalog No. p-7077186
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC183391000 100 g
AC183390010 1 kg
AC183390025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC183391000 Supplier Thermo Scientific Chemicals Supplier No. 183391000
Only null left

Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Melting Point 230.0°C to 240.0°C
pH 4.5 to 6.5 (4% aq. soln.)
Infrared Spectrum Authentic
Linear Formula [-CH2CH(OH)-]n
Quantity 100 g
Merck Index 15,7706
Solubility Information Solubility in water: soluble in hot water.
Viscosity 27-31 mPa.s (20°C 4% aq. sol.)
Percent Purity 95.5-96.5%
Chemical Name or Material Poly(vinyl alcohol)

RUO – Research Use Only