missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000

Catalog No. AC184965000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184965000 500 g Glass bottle
AC184960250 25 g Glass bottle
AC184961000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC184965000 Supplier Thermo Scientific Chemicals Supplier No. 184965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27119-07-9 | (C7H13NO4S)n | 207.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27119-07-9 , 7732-18-5
Molecular Formula (C7H13NO4S)n
Molecular Weight (g/mol) 207.24
MDL Number MFCD00084369
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*

Specifications

Physical Form Viscous Liquid
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
Percent Purity 8.0 to 12.0% (solids)
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only