accessibility menu, dialog, popup

UserName

∗NHOSOOHO∗MeMen

CAS RN 27119-07-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C7H13NO4S)n
InChi Key: —
SMILES: CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)