missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PNPP Substrates

Catalog No. PI34045
Change view
Click to view available options
Format:
Kit
Liqiuid
Powder
Tablet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
PI34045 Powder
PI34047 Tablet
PI37620 Kit
PI37621 Liqiuid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI34045 Supplier Thermo Scientific™ Supplier No. 34045

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.

PNPP (p-Nitrophenyl Phosphate, Disodium Sal) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution, and a complete kit containing substrate solution and diethanolamine buffer.

Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.

Features of PNPP (p-nitrophenyl phosphate):

  • One-component alkaline phosphatase (AP) substrate
  • Absorbance maximum of 405 nm
  • Develops a yellow hue in solution upon reaction with AP
  • Safer to use—contains a non-mercury, non-azide preservative

The substrate is offered in several formats:

PNPP Substrate

  • Powder Form

PNPP Substrate Tablets

  • 5 mg tablets with an inert filler as the balance

PNPP Substrate Kit

  • PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.

1-Step™ PNPP Substrate Solution

  • Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.

Specifications

Description PNPP Powder
Product Type ELISA Substrate
Quantity 25 g
Detection Method Colorimetric
Form Powder
Format Powder
Substrate Properties Chemical Substrate
Substrate Type Alkaline Phosphatase
Content And Storage Upon receipt store product at 4°C.
Shipping Condition Ambient
What alkaline phosphatase substrates do you offer?