missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PlusOne™ Reagents
High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.
$114.28 - $444.76
Description
Urea
- Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
- Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
- 8M urea included in the 2-D sample solution
Low toxicity and environmentally safe solvent
- Used for covalently attach polyacrylamide gels to a glass surface
- Firmly attaches to the glass during staining and drying procedures
Urea:
- Molecular Weight: 60.06
- Physical state: Solid
- Color: White
- Pack of 500g
- Molecular Weight: 129.06
- Physical state: Liquid
- Color: Colorless
- Pack of 500mL
- Molecular Weight: 248.4
- Physical state: Liquid
- Color: Pale
- Pack of 25mL
IEF, PAGE, sequencing
Certifications
Suitable for DIGE
Specifications
| 2530-85-0 |
Safety and Handling
ShelfLife : 12 Months