missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlusOne™ Reagents

Catalog No. 45000244
Change view
Click to view available options
Quantity:
500 g
500 mL
Form:
Liquid
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
45000244 500 mL Liquid
45000234 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000244 Supplier Cytiva Supplier No. 17133201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.

Urea

  • Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
  • Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
  • 8M urea included in the 2-D sample solution
Repel-Silane ES
Low toxicity and environmentally safe solvent
  • Inhibits sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces
  • Bind-Silane
    • Used for covalently attach polyacrylamide gels to a glass surface
    • Firmly attaches to the glass during staining and drying procedures

    Urea:

    • Molecular Weight: 60.06
    • Physical state: Solid
    • Color: White
    • Pack of 500g
    Repel-Silane ES
    • Molecular Weight: 129.06
    • Physical state: Liquid
    • Color: Colorless
    • Pack of 500mL
    Bind-Silane
    • Molecular Weight: 248.4
    • Physical state: Liquid
    • Color: Pale
    • Pack of 25mL

    IEF, PAGE, sequencing

    Certifications

    Suitable for DIGE

    Specifications

    CAS 2530-85-0
    Description PlusOne Repel-Silane ES
    Color Colorless
    Concentration 2%
    Form Liquid
    For Use With (Application) Used in IEF, PAGE and Sequencing
    Molecular Weight (g/mol) 129.06
    Quantity 500 mL
    Storage Requirements Dry and dark at room temperature
    Product Type Urea