missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments

Catalog No. p-4525907
Change view
Click to view available options
Needle Gauge:
23 G
26 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
365A0241 23 G
03170518 26 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 365A0241 Supplier Thermo Scientific™ Supplier No. 365A0241
Only null left

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
  • Specifications

    Description 0.5 μL volume; 23 gauge; 57 mm length
    Needle Gauge 23 G
    Length (Metric) 57 mm
    Product Type GC Autosampler Syringe, Removable Needle
    Disposable Reusable
    Quantity Ea.
    Volume (Metric) 0.5 μL
    Length (Metric) Needle 57 mm
    Needle Point Style Cone
    For Use With (Equipment) TriPlus RSH Autosampler and TriPlus 100LS Autosampler
    Connection Type Removable
    Show More Show Less