missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PLpro inhibitor 6

Catalog No. 735725
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
735725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 735725 Supplier Tocris Bioscience™ Supplier No. 7357/25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SARS-CoV and SARS-CoV-2 PLpro inhibitor

PLpro inhibitor 6 is a SARS-CoV and SAR-CoV-2 PLpro inhibitor (IC50 values are 2.6 and 5.0 μM, respectively). Inhibits SARS-CoV and SARS-CoV-2 infection of Vero E6 cells in vitro (EC50 values are 13.1 and 21 μM, respectively).

Specifications

Chemical Name or Material 5-(Acetylamino)-2-methyl-N-[(1 R)-1-(1-naphthalenyl)ethyl]benzamide
CAS 1093070-14-4
Quantity 25 mg
Target Papain-like Protease Inhibitors
Molecular Formula C22H22N2O2
Purity 0.98