missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, DNA-free

Catalog No. p-8739012
Encompass
Change view
Click to view available options
No. of Reactions:
2000 Reactions
400 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
15966005 400 Reactions
15966025 2000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15966005 Supplier Invitrogen™ Supplier No. 15966005
Only null left

Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA.

Invitrogen Platinum Taq DNA Polymerase, DNA-free, is manufactured using a novel, closed single-use system together with stringent quality-control testing to minimize the risk of contamination with bacterial and human DNA. This enzyme is an ideal choice for PCR-based applications requiring the high sensitivity without false-positive results from reagent-borne contamination. This DNA-free enzyme provides the same high level of performance and lot-to-lot consistency as standard Platinum Taq DNA Polymerase.

Platinum Taq DNA Polymerase, DNA-free, is a recombinant Taq polymerase complexed with a proprietary antibody that blocks polymerase activity at ambient temperatures and dissociates after the initial denaturation step at 94°C. This automatic hot start provides increased sensitivity, specificity, and yield, while allowing convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase, DNA-free, has a non-template–dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Features

  • Certified low DNA contamination level (≤0.01 copy of bacterial gDNA per enzyme unit)
  • Confidence in qualitative and quantitative detection of low-abundance microorganisms due to signal absence with no-template/reagents-only PCR controls
  • Same performance as Platinum Taq DNA Polymerase manufactured using conventional process

Applications

  • Amplification of DNA from complex genomic and viral templates
  • Hot start PCR
  • RT-PCR

Specifications

Concentration 5 U/μL
Content And Storage • Platinum Taq DNA Polymerase, DNA-free, 100 μL
• 10X PCR Buffer (without magnesium), DNA-free, 1.25 mL
• 50 mM MgCl2, DNA-free (1 mL)

Store at -20°C in a non-frost-free freezer.
Format Tube
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Product Type Taq DNA Polymerase
Quantity 500 Units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 400 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less
What is the unit concentration of Platinum Taq DNA Polymerase, DNA-free?

Platinum Taq DNA Polymerase, DNA-free is available in 5.0 U/µL enzyme concentration.

Can I assemble my PCR reactions with Platinum Taq DNA Polymerase, DNA-free at room temperature?

Yes. Due to stable antibody-mediated hot-start technology, Platinum Taq DNA Polymerase, DNA-free is highly stable. The PCR reactions can be assembled at room temperature without any loss of amplification specificity.

I am using Platinum Taq DNA Polymerase, DNA-free for PCR. "No-template controls" in my experiment are showing a background amplification signal. What should I do differently?

Amplification signal in “no-template controls” (NTC), also known as false-positive signal, or background amplification in “reagent-only control”, may arise from contaminating DNA that entered control reactions from the environment, from the researcher, or was introduced into the PCR via contaminated PCR reagents or consumables. To avoid contaminating DNA in your experiments, it is important to follow these recommendations:

- Be cautious to maintain a DNA-free environment during handling and opening the vials with DNA polymerase, PCR buffer, and MgCl2
- Ensure that all components of the PCR, like dNTPs, primers, probes, or water are free of contaminating DNA that can give false positive PCR results
- Avoid opening the tubes containing DNA-free reagents multiple times to minimize risk of DNA contamination
- Work in a UV-irradiated workstation
- Use certified DNA-free pipette tips and PCR consumables

Can I use the same cycling protocol that I use with standard Taq for PCR reactions with Platinum Taq DNA Polymerase, DNA-free?

PCR reactions with Platinum Taq DNA Polymerase, DNA-free can be run using the same cycling protocol as that for standard Taq.

Can I use Platinum Taq DNA Polymerase, DNA-free in qPCR?

Yes, Platinum Taq DNA Polymerase, DNA-free can be used in qPCR master mixes for target detection and quantification in a real-time PCR instrument using probes or SYBR Green dye.

What other PCR components do you recommend using with Platinum Taq DNA Polymerase, DNA-free?

To avoid false positive results, we recommend using Platinum Taq DNA Polymerase, DNA-free with 10 mM dNTP Mix (Cat. No. 18427088) and RT-PCR Grade Water (Cat. No. AM9935).

What are the purity requirements for Platinum Taq DNA Polymerase, DNA-free?

Platinum Taq DNA Polymerase, DNA-free is certified to contain low-DNA contamination level: ≤0.01 copy of bacterial gDNA per enzyme unit; ≤0.001 copy/enzyme unit of human gDNA; ≤0.01 copy/enzyme unit of plasmid DNA.

What is the difference between Platinum Taq DNA Polymerase and Platinum Taq DNA Polymerase, DNA-free?

Platinum Taq DNA Polymerase, DNA-free is manufactured using closed and single-use system technology to minimize DNA contamination risk. Platinum Taq DNA Polymerase, DNA-free provides the same high level of performance and lot-to-lot consistency that is expected for the standard Platinum Taq DNA Polymerase.


For Research Use Only. Not for use in diagnostic procedures.