Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
$2,708.38 - $3,644.25
Specifications
| Material | Polystyrene |
|---|---|
| No. per Case | 30 |
| No. per Pack | 5 |
| Plate Blocking | Unspecified |
| Target Molecule | Peptides |
| Catalog Number | Mfr. No. | Color | Volume (Metric) Working | Product Type | Surface Treatment | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Volume (Metric) Working | Product Type | Surface Treatment | Price | Quantity & Availability | ||
|
1256712
|
Thermo Scientific™
436006 |
Clear | 100 μL | Microplate | Covalent Binding |
Case of 30 Each for $2,708.38
|
|
||
|
1256710
|
Thermo Scientific™
436007 |
White | 100 μL | Solid plate 96-Well | Covalent Binding |
Case of 30 Each for $3,012.63
|
|
||
|
1256711
|
Thermo Scientific™
436008 |
Black | 100 μL | Microplate | Amine |
Case of 30 Each for $3,012.63
|
|
||
|
1256713
|
Thermo Scientific™
436013 |
Clear | 100 μL | Microplate | Amine |
Case of 30 Each for $3,644.25
|
|
||
|
1256714
|
Thermo Scientific™
436023 |
Clear | 100 μL | Microplate | Amine |
Case of 30 Each for $3,644.25
|
|
||
|
12565132
|
Thermo Scientific™
478042 |
Clear | 100 μL | Solid Module F8 (x12) in Frame | Covalent Binding |
Case of 30 Each for $3,312.94
|
|
||
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
- Minimal leaching
- Withstands vigorous washing
- Coating with lower amounts of reagent may be possible
- Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
| Polystyrene | |
| 5 | |
| Peptides |
| 30 | |
| Unspecified |
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.