missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces

Catalog No. 1256711
Change view
Click to view available options
Color:
Black
Clear
White
Volume (Metric) Working:
100 μL
Product Type:
Microplate
Solid Module F8 (x12) in Frame
Solid plate 96-Well
Surface Treatment:
Amine
Covalent Binding
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Working Product Type Surface Treatment
1256711 Black 100 μL Microplate Amine
1256712 Clear 100 μL Microplate Covalent Binding
1256710 White 100 μL Solid plate 96-Well Covalent Binding
1256713 Clear 100 μL Microplate Amine
1256714 Clear 100 μL Microplate Amine
12565132 Clear 100 μL Solid Module F8 (x12) in Frame Covalent Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 1256711 Supplier Thermo Scientific™ Supplier No. 436008
Only null left

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

Specifications

Format Standard
Detection Method Fluorescence
Array 8 x 12
Packaging 30 Cases
Binding Type Covalent
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment Amine
Total Volume (Metric) Well 400 μL
Well Shape Round, Flat Bottom (F-well)
No. per Case 30
No. per Pack 5
Plate Blocking Unspecified
Target Molecule Peptides
Show More Show Less

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.