Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Supplier:  Thermo Scientific™ 436013
 View more versions of this product
Catalog No. 1256713
Only null left
Description
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Chemical Identifiers
Polystyrene
5
Peptides
30
Unspecified
Specifications
Colorimetric
Covalent
Microplate
100 μL
Amine
30
Unspecified
30 Cases
Clear
Polystyrene
Non-sterile
Round, Flat Bottom (F-well)
5
Peptides
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.