missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pitavastatin calcium

Catalog No. 494210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
494210 10 mg
494250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 494210 Supplier Tocris Bioscience™ Supplier No. 4942/10

HMG-CoA reductase inhibitor

Synthetic inhibitor of HMG-CoA reductase (Ki = 1.7 nM). Lowers low-density lipoprotein (LDL) cholesterol levels. Displays minimal metabolism by cytochrome P450 enzymes, with little involvement of CYP3A4. Shown to improve cardiac function and survival in a rat model of hypertensive heart failure.

Chemical Identifiers

CAS 147526-32-7
Molecular Formula C50H46CaF2N2O8

Specifications

Quantity 10 mg
Formula Weight 440.5
Purity >99%
Molecular Weight (g/mol) 881.00
Chemical Name or Material Pitavastatin calcium
Recommended Storage Desiccate at Room Temperature