missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroctone Olamine 97.0+%, TCI America™
$262.83 - $877.97
Chemical Identifiers
| CAS | 68890-66-4 |
|---|---|
| Molecular Formula | C16H30N2O3 |
| Molecular Weight (g/mol) | 298.427 |
| MDL Number | MFCD01690792 |
| InChI Key | BTSZTGGZJQFALU-UHFFFAOYSA-N |
| Synonym | 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine |
| PubChem CID | 50258 |
| IUPAC Name | 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one |
| SMILES | CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N |
Chemical Identifiers
| 68890-66-4 | |
| 298.427 | |
| BTSZTGGZJQFALU-UHFFFAOYSA-N | |
| 50258 | |
| CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N |
| C16H30N2O3 | |
| MFCD01690792 | |
| 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine | |
| 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one |
Specifications
| 68890-66-4 | |
| White | |
| MFCD01690792 | |
| 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine | |
| CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N | |
| 298.427 | |
| 298.43 | |
| Crystalline Powder |
| 136°C | |
| C16H30N2O3 | |
| 1 g | |
| BTSZTGGZJQFALU-UHFFFAOYSA-N | |
| 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one | |
| 50258 | |
| ≥97.0% (HPLC,T) | |
| Piroctone Olamine |
Safety and Handling
RTECSNumber : UU7786150
TSCA : No
Recommended Storage : Refrigerator