missing translation for 'onlineSavingsMsg'
Learn More

Piroctone Olamine 97.0+%, TCI America™

Supplier:  TCI America P21781G

 View more versions of this product

Catalog No. P21781G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Piroctone Olamine
136°C
C16H30N2O3
1 g
BTSZTGGZJQFALU-UHFFFAOYSA-N
2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one
50258
≥97.0% (HPLC,T)
68890-66-4
White
MFCD01690792
1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine
CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N
298.427
298.43
Crystalline Powder

Chemical Identifiers

68890-66-4
298.427
BTSZTGGZJQFALU-UHFFFAOYSA-N
50258
CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N
C16H30N2O3
MFCD01690792
1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine
2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UU7786150

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions