missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroctone Olamine 97.0+%, TCI America™
Supplier: TCI America P21785G
Specifications
| Piroctone Olamine | |
| 136°C | |
| C16H30N2O3 | |
| 5 g | |
| BTSZTGGZJQFALU-UHFFFAOYSA-N | |
| 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one | |
| 50258 | |
| ≥97.0% (HPLC,T) |
| 68890-66-4 | |
| White | |
| MFCD01690792 | |
| 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine | |
| CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N | |
| 298.427 | |
| 298.43 | |
| Crystalline Powder |
Chemical Identifiers
| 68890-66-4 | |
| 298.427 | |
| BTSZTGGZJQFALU-UHFFFAOYSA-N | |
| 50258 | |
| CC1=CC(=O)N(C(=C1)CC(C)CC(C)(C)C)O.C(CO)N |
| C16H30N2O3 | |
| MFCD01690792 | |
| 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine | |
| 2-aminoethanol;1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2-one |
Safety and Handling
RTECSNumber : UU7786150
TSCA : No
Recommended Storage : Refrigerator